Gene Transcripts

Gene Symbol
BUD13
Entrez Gene ID
84811
Gene Name
BUD13 homolog
Gene Aliases
Cwc26, fSAP71
Chromosome Location
Chr. 11: 116748170 - 116772998 on Build GRCh38
UniGene ID
Hs.437341
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159736.1 NP_001153208.1 4 390
NM_032725.3 NP_116114.1 4 390
XM_011543035.2 XP_011541337.1 4 964
GenBank mRNA AK057832.1 - 4 379
AK225164.1 - 4 379
AK289635.1 - 4 379
AK291111.1 - 4 379
AK311068.1 - 4 379
BC006350.2 - 4 380 4111572
DA042987.1 - 4 391

View Full Product Details