Gene Transcripts

Gene Symbol
SPSB3
Entrez Gene ID
90864
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
C16orf31, SSB3
Chromosome Location
Chr. 16: 1776712 - 1782605 on Build GRCh38
UniGene ID
Hs.592080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324081.1 NP_001311010.1 7 900
NM_080861.3 NP_543137.2 7 922
XM_005255673.4 XP_005255730.1 7 1112
GenBank mRNA AF403028.1 - 6 831
AK057850.1 - 680
AK074190.1 - 7 886
BC007588.1 - 7 868 3342825
BC024244.1 - 7 886 4798873
BC035065.1 - 7 879
BC041897.1 - 6 1696 5298729
BC047441.1 - 7 864 5303706
BC065515.1 - 4 1165
BQ930544.1 - 2 204

View Full Product Details