Gene Transcripts

Gene Symbol
CCDC74B
Entrez Gene ID
91409
Gene Name
coiled-coil domain containing 74B
Gene Aliases
Chromosome Location
Chr. 2: 130139283 - 130145134 on Build GRCh38
UniGene ID
Hs.29383
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258307.1 NP_001245236.1 4 524
NM_207310.2 NP_997193.1 4 722
XM_005263840.2 XP_005263897.1 4 747
XM_005263842.2 XP_005263899.1 4 549
XM_006712833.2 XP_006712896.1 4 759
XM_006712834.3 XP_006712897.1 4 699
XM_006712835.2 XP_006712898.1 4 561
XM_006712837.2 XP_006712900.1 4 501
XM_011512142.2 XP_011510444.1 2 990
GenBank mRNA AK057609.1 - 4 646
AK297246.1 - 3 1634
AL133619.1 - 1 1772
BC067771.1 - 4 469 4824929

Gene Symbol
CCDC74A
Entrez Gene ID
90557
Gene Name
coiled-coil domain containing 74A
Gene Aliases
Chromosome Location
Chr. 2: 131522621 - 131533666 on Build GRCh38
UniGene ID
Hs.351461
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258304.1 NP_001245233.1 4 847
NM_001258305.1 NP_001245234.1 4 721
NM_001258306.1 NP_001245235.1 4 523
NM_138770.2 NP_620125.1 4 721
XM_005263835.3 XP_005263892.1 4 621
XM_005263836.4 XP_005263893.1 2 1004
XM_005263837.1 XP_005263894.1 4 639
XM_006712828.3 XP_006712891.1 4 820
XM_017005234.1 XP_016860723.1 4 833
XM_017005235.1 XP_016860724.1 4 892
XM_017005236.1 XP_016860725.1 4 634
XM_017005237.1 XP_016860726.1 4 848
XM_017005238.1 XP_016860727.1 4 857
XM_017005239.1 XP_016860728.1 4 694
XM_017005240.1 XP_016860729.1 4 880
XM_017005241.1 XP_016860730.1 5 629
XM_017005242.1 XP_016860731.1 4 630
XM_017005243.1 XP_016860732.1 4 641
XM_017005244.1 XP_016860733.1 5 425
XM_017005245.1 XP_016860734.1 4 651
XM_017005246.1 XP_016860735.1 4 523
GenBank mRNA AB299086.1 - 4 762
AB299087.1 - 4 636
AK097664.1 - 2 1204
BC016861.1 - 4 636 3914851
BC063387.1 - 4 460 5494806
CV023784.1 - 4 584
DA375407.1 - 4 523

View Full Product Details