Gene Transcripts

Gene Symbol
LENG8
Entrez Gene ID
114823
Gene Name
leukocyte receptor cluster member 8
Gene Aliases
pp13842
Chromosome Location
Chr. 19: 54448719 - 54462046 on Build GRCh38
UniGene ID
Hs.502378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052925.3 NP_443157.1 4 510
XM_005278248.4 XP_005278305.1 4 501
XM_005278249.4 XP_005278306.1 4 953
XM_005278250.4 XP_005278307.1 3 372
XM_006722996.3 XP_006723059.1 3 377
XM_011526413.2 XP_011524715.1 4 440
XM_011526415.2 XP_011524717.1 4 501
GenBank mRNA BC028048.1 - 4 461
CN403513.1 - 4 450

View Full Product Details