Gene Transcripts

Gene Symbol
OR8I2
Entrez Gene ID
120586
Gene Name
olfactory receptor family 8 subfamily I member 2
Gene Aliases
OR11-170
Chromosome Location
Chr. 11: 56093308 - 56094240 on Build GRCh38
UniGene ID
Hs.554524
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003750.1 NP_001003750.1 1 763
GenBank mRNA BC137007.1 - 1 794
BC137008.1 - 1 794 9021001

View Full Product Details