Gene Transcripts

Gene Symbol
OR10A7
Entrez Gene ID
121364
Gene Name
olfactory receptor family 10 subfamily A member 7
Gene Aliases
OR12-6
Chromosome Location
Chr. 12: 55221025 - 55221975 on Build GRCh38
UniGene ID
Hs.554557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005280.1 NP_001005280.1 1 659

View Full Product Details