Gene Transcripts

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
Chromosome Location
Chr. 17: 18950676 - 19020691 on Build GRCh38
UniGene ID
Hs.462418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042450.2 NP_001035915.1 13, 14 1665
NM_001270648.1 NP_001257577.1 12, 13 1584
NM_001270649.1 NP_001257578.1 12, 13 1557
NM_001282417.1 NP_001269346.1 13, 14 1774
NM_152351.4 NP_689564.3 13, 14 1713
XM_011523652.1 XP_011521954.1 14, 15 1932
XM_011523653.2 XP_011521955.1 13, 14 1904
XM_017024191.1 XP_016879680.1 14, 15 1981
GenBank mRNA AK057946.1 - 13, 14 1714
AK298345.1 - 12, 13 1543
BC034380.1 - 13, 14 1610
BC039868.1 - 13, 14 1791 4157703
BC062617.1 - 12, 13 1538

View Full Product Details