Gene Transcripts

Gene Symbol
H1FOO
Entrez Gene ID
132243
Gene Name
H1 histone family member O, oocyte specific
Gene Aliases
H1.8, H1oo, osH1
Chromosome Location
Chr. 3: 129543214 - 129555185 on Build GRCh38
UniGene ID
Hs.97358
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308262.1 NP_001295191.1 2 304
NM_153833.2 NP_722575.1 3 595
XM_017005733.1 XP_016861222.1 2 341
GenBank mRNA AY158091.1 - 3 595
BC047943.1 - 2 303 5742122
BM564157.1 - 321

View Full Product Details