Gene Transcripts
- Gene Symbol
- STXBP5
- Entrez Gene ID
- 134957
- Gene Name
- syntaxin binding protein 5
- Gene Aliases
- LGL3, LLGL3, Nbla04300
- Chromosome Location
- Chr. 6: 147204184 - 147390476 on Build GRCh38
- UniGene ID
- Hs.736439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127715.2 |
NP_001121187.1
|
15, 16 |
1806 |
|
NM_139244.4 |
NP_640337.3
|
15, 16 |
1806 |
|
XM_006715339.2 |
XP_006715402.1
|
15, 16 |
1980 |
|
XM_011535453.1 |
XP_011533755.1
|
15, 16 |
1578 |
|
XM_017010267.1 |
XP_016865756.1
|
10, 11 |
1877 |
|
XM_017010268.1 |
XP_016865757.1
|
15, 16 |
1843 |
|
GenBank mRNA |
AB073398.1 |
- |
6, 7 |
645 |
|
AK090549.1 |
- |
5, 6 |
516 |
|
AK128121.1 |
- |
15, 16 |
1573 |
|
AL834152.1 |
- |
15, 16 |
1792 |
|
BC113382.1 |
- |
15, 16 |
1679 |
8322434 |
BC113408.1 |
- |
15, 16 |
1679 |
8322460 |
View Full Product Details