Gene Transcripts

Gene Symbol
SMN2
Entrez Gene ID
6607
Gene Name
survival of motor neuron 2, centromeric
Gene Aliases
BCD541, C-BCD541, GEMIN1, SMNC, TDRD16B
Chromosome Location
Chr. 5: 70049523 - 70077595 on Build GRCh38
UniGene ID
Hs.202179
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017411.3 NP_059107.1 4 506
NM_022875.2 NP_075013.1 4 506
NM_022876.2 NP_075014.1 4 506
NM_022877.2 NP_075015.1 4 506
XM_011543599.1 XP_011541901.1 4 385
XM_017009787.1 XP_016865276.1 4 385
GenBank mRNA AK289669.1 - 4 389
BC000908.2 - 4 375 2985046
BC015308.1 - 4 373 4427000
BC070242.1 - 4 352 4673318
CN272420.1 - 2 209
JQ690864.1 - 2 95
JQ690865.1 - 2 95
JQ732167.1 - 2 95
U21914.1 - 4 340

Gene Symbol
SMN1
Entrez Gene ID
6606
Gene Name
survival of motor neuron 1, telomeric
Gene Aliases
BCD541, GEMIN1, SMA, SMA1, SMA2, SMA3, SMA4, SMA@, SMN, SMNT, T-BCD541, TDRD16A
Chromosome Location
Chr. 5: 70924941 - 70953015 on Build GRCh38
UniGene ID
Hs.535788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000344.3 NP_000335.1 4 506
NM_001297715.1 NP_001284644.1 4 506
NM_022874.2 NP_075012.1 4 506
XM_011543596.1 XP_011541898.1 4 385
XM_017009786.1 XP_016865275.1 4 385
GenBank mRNA BC062723.1 - 4 362
JQ657798.1 - 2 95
JQ657799.1 - 2 95
JQ657801.1 - 2 95
JQ657802.1 - 2 95
JQ732166.1 - 2 95
U18423.1 - 4 376

View Full Product Details