Gene Transcripts

Gene Symbol
ZNRF4
Entrez Gene ID
148066
Gene Name
zinc and ring finger 4
Gene Aliases
RNF204, SPERIZIN, Ssrzf1, spzn
Chromosome Location
Chr. 19: 5455415 - 5456856 on Build GRCh38
UniGene ID
Hs.126496
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181710.3 NP_859061.3 1 163
GenBank mRNA AK292251.1 - 1 147
BC017592.2 - 1 147 4828408

View Full Product Details