Gene Transcripts

Gene Symbol
SLC9B1
Entrez Gene ID
150159
Gene Name
solute carrier family 9 member B1
Gene Aliases
NHA1, NHEDC1
Chromosome Location
Chr. 4: 102885048 - 103019739 on Build GRCh38
UniGene ID
Hs.666728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100874.2 NP_001094344.1 4 395
NM_139173.3 NP_631912.2 4 395
NR_047513.1 4 395
XM_006714093.3 XP_006714156.1 5 414
XM_011531622.1 XP_011529924.1 5 378
XM_011531623.1 XP_011529925.1 4 382
GenBank mRNA AF447585.1 - 4 375
AK313592.1 - 4 322
AY461581.1 - 4 343
BC046636.1 - 4 319
BC110794.1 - 4 279 5745227
BC132712.1 - 4 322 40146734
BC136966.1 - 4 322 9020958
DQ003059.1 - 4 375

View Full Product Details