Gene Transcripts

Gene Symbol
NFAM1
Entrez Gene ID
150372
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
CNAIP
Chromosome Location
Chr. 22: 42380407 - 42438155 on Build GRCh38
UniGene ID
Hs.436677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318323.1 NP_001305252.1 2 283
NM_145912.6 NP_666017.1 2 283
XM_005261374.4 XP_005261431.1 4 910
GenBank mRNA AY121370.1 - 2 283
AY247409.1 - 2 278
BC038241.1 - 2 278 5180790
BI906855.1 - 2 257
CT841509.1 - 2 283

View Full Product Details