Gene Transcripts

Gene Symbol
C2orf15
Entrez Gene ID
150590
Gene Name
chromosome 2 open reading frame 15
Gene Aliases
Chromosome Location
Chr. 2: 99141613 - 99151487 on Build GRCh38
UniGene ID
Hs.352211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317992.1 NP_001304921.1 5 656
NM_144706.3 NP_653307.1 4 560
GenBank mRNA AK314133.1 - 4 560
BC021264.2 - 4 451 4542657
BX364103.2 - 5 518
DA396675.1 - 4 560

View Full Product Details