Gene Transcripts

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
Chromosome Location
Chr. 5: 132750817 - 132778216 on Build GRCh38
UniGene ID
Hs.522057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098811.1 NP_001092281.1 10 1760
XM_005271927.2 XP_005271984.1 10 1759
XM_005271928.2 XP_005271985.1 10 1756
XM_005271931.3 XP_005271988.1 11 1843
XM_011543288.1 XP_011541590.1 10 1782
XM_017009254.1 XP_016864743.1 10 1753
XM_017009255.1 XP_016864744.1 10 1826
XM_017009256.1 XP_016864745.1 10 1823
XM_017009257.1 XP_016864746.1 10 1776
GenBank mRNA AF440762.1 - 10 1760

View Full Product Details