Gene Transcripts

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
Chromosome Location
Chr. 5: 132750817 - 132778216 on Build GRCh38
UniGene ID
Hs.522057
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098812.1 NP_001092282.1 10 1966
NM_001098813.1 NP_001092283.1 10 2160
NM_001300798.1 NP_001287727.1 10 1960
NM_001300799.1 NP_001287728.1 10 2228
NM_015146.1 NP_055961.1 10 2234
XM_017009258.1 XP_016864747.1 10 2035
XM_017009259.1 XP_016864748.1 10 2029
XM_017009260.1 XP_016864749.1 10 1982
XM_017009261.1 XP_016864750.1 10 2297
GenBank mRNA AF179995.1 - 10 2234
BC140759.1 - 10 2050 9021627
BC171779.1 - 10 2051 9054433
D86957.1 - 10 2234

View Full Product Details