Gene Transcripts

Gene Symbol
SIGLEC14
Entrez Gene ID
100049587
Gene Name
sialic acid binding Ig like lectin 14
Gene Aliases
Chromosome Location
Chr. 19: 51639478 - 51646879 on Build GRCh38
UniGene ID
Hs.707374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098612.1 NP_001092082.1 7 2052
XM_017026113.1 XP_016881602.1 6 1740
GenBank mRNA AY854038.1 - 7 1976
CA439587.1 - 1 267

View Full Product Details