Gene Transcripts

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
Chromosome Location
Chr. 6: 31114831 - 31140092 on Build GRCh38
UniGene ID
Hs.310958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014068.2 NP_054787.2 6 825
GenBank mRNA AB031479.1 - 6 810
AF484418.1 - 6 825
AF484419.1 - 2 430
BI825917.1 - 3 487

View Full Product Details