Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278562.1 | NP_001265491.1 | 8 | 889 | |
NM_207338.3 | NP_997221.2 | 9 | 1180 | ||
NR_103739.1 | 7 | 902 | |||
XM_017021999.1 | XP_016877488.1 | 9 | 1099 | ||
GenBank mRNA | AK090598.1 | - | 8 | 874 | |
AK311441.1 | - | 7 | 902 | ||
AY358729.1 | - | 9 | 1180 |