Gene Transcripts

Gene Symbol
KLHDC8B
Entrez Gene ID
200942
Gene Name
kelch domain containing 8B
Gene Aliases
CHL
Chromosome Location
Chr. 3: 49171120 - 49176487 on Build GRCh38
UniGene ID
Hs.13781
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173546.2 NP_775817.1 4 968
XM_005264938.2 XP_005264995.1 4 1454
XM_005264940.4 XP_005264997.1 4 1089
XM_006713015.2 XP_006713078.1 4 1484
XM_006713016.2 XP_006713079.1 4 1463
GenBank mRNA AK002164.1 - 2 208
AK294160.1 - 4 817
AY129011.1 - 4 925
BC039083.1 - 4 918 5167173

View Full Product Details