Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
201191
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
Chromosome Location
Chr. 17: 50111309 - 50129882 on Build GRCh38
UniGene ID
Hs.567769
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174920.3 NP_777580.1 8, 9 1287
XM_017024324.1 XP_016879813.1 8, 9 2142
GenBank mRNA AY148482.1 - 8, 9 1208

View Full Product Details