Gene Transcripts
- Gene Symbol
- ADCK5
- Entrez Gene ID
- 203054
- Gene Name
- aarF domain containing kinase 5
- Gene Aliases
- Chromosome Location
- Chr. 8: 144373762 - 144393242 on Build GRCh38
- UniGene ID
- Hs.283374
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_174922.4 |
NP_777582.4
|
9, 10 |
1095 |
|
XM_006716527.2 |
XP_006716590.1
|
9, 10 |
1085 |
|
XM_011516907.2 |
XP_011515209.1
|
9, 10 |
1808 |
|
XM_011516909.1 |
XP_011515211.1
|
9, 10 |
1095 |
|
XM_011516910.1 |
XP_011515212.1
|
10, 11 |
1118 |
|
XM_017013174.1 |
XP_016868663.1
|
9, 10 |
1133 |
|
GenBank mRNA |
AK092773.1 |
- |
9, 10 |
1058 |
|
BC085013.1 |
- |
4, 5 |
405 |
|
BC101659.1 |
- |
9, 10 |
1035 |
8069165 |
BC143326.1 |
- |
8, 9 |
862 |
|
BU845476.1 |
- |
4, 5 |
403 |
|
View Full Product Details