Gene Transcripts

Gene Symbol
SUSD3
Entrez Gene ID
203328
Gene Name
sushi domain containing 3
Gene Aliases
Chromosome Location
Chr. 9: 93058688 - 93085138 on Build GRCh38
UniGene ID
Hs.88417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287005.1 NP_001273934.1 6 1228
NM_001287006.1 NP_001273935.1 4 982
NM_001287007.1 NP_001273936.1 3 793
NM_001287008.1 NP_001273937.1 4 925
NM_145006.3 NP_659443.1 5 1114
XM_011518358.1 XP_011516660.1 8 2308
XM_011518359.1 XP_011516661.1 7 2258
XM_011518361.2 XP_011516663.1 8 1370
XM_011518362.1 XP_011516664.1 6 2144
XM_017014448.1 XP_016869937.1 6 1415
XM_017014449.1 XP_016869938.1 5 1296
XM_017014450.1 XP_016869939.1 7 1255
XM_017014451.1 XP_016869940.1 5 1864
GenBank mRNA AK128289.1 - 3 3082
AY358190.1 - 6 1156
BC014601.2 - 5 1095 4821517
BC041834.1 - 3 3008 5268318
BM458621.1 - 3 671

View Full Product Details