Gene Transcripts

Gene Symbol
CCZ1B
Entrez Gene ID
221960
Gene Name
CCZ1 homolog B, vacuolar protein trafficking and biogenesis associated
Gene Aliases
C7orf28B, H_NH0577018.2
Chromosome Location
Chr. 7: 6798935 - 6826295 on Build GRCh38
UniGene ID
Hs.567779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198097.3 NP_932765.1 6 598
GenBank mRNA AK292326.1 - 6 566
AK296026.1 - 5 404
AL050215.1 - 4 240
BC010130.2 - 6 533 3953739

Gene Symbol
CCZ1
Entrez Gene ID
51622
Gene Name
CCZ1 homolog, vacuolar protein trafficking and biogenesis associated
Gene Aliases
C7orf28A, CCZ1A, CGI-43, H_DJ1163J12.2
Chromosome Location
Chr. 7: 5898710 - 5925974 on Build GRCh38
UniGene ID
Hs.530000
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015622.5 NP_056437.4 6 590
GenBank mRNA AF151801.1 - 6 634
BC132749.1 - 6 575 40146771
BC132751.1 - 6 575 40146773
BI546694.1 - 6 582
CN310516.1 - 3 282

View Full Product Details