Gene Transcripts
- Gene Symbol
- HCG27
- Entrez Gene ID
- 253018
- Gene Name
- HLA complex group 27 (non-protein coding)
- Gene Aliases
- bCX101P6.9, bPG299F13.9, bQB115I13.2
- Chromosome Location
- Chr. 6: 31197760 - 31203968 on Build GRCh38
- UniGene ID
- Hs.659818
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026791.1 |
|
2 |
1017 |
|
GenBank mRNA |
AK097442.1 |
- |
2 |
1017 |
|
BC041465.2 |
- |
1 |
896 |
|
View Full Product Details