Gene Transcripts

Gene Symbol
KRTAP15-1
Entrez Gene ID
254950
Gene Name
keratin associated protein 15-1
Gene Aliases
KAP15.1
Chromosome Location
Chr. 21: 30440328 - 30440780 on Build GRCh38
UniGene ID
Hs.553654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181623.1 NP_853654.1 1 72
GenBank mRNA AB096942.1 - 1 94
BC104925.1 - 1 121 8143928
BC104927.1 - 1 121 8143930

View Full Product Details