Gene Transcripts

Gene Symbol
SYT14
Entrez Gene ID
255928
Gene Name
synaptotagmin 14
Gene Aliases
SCAR11, sytXIV
Chromosome Location
Chr. 1: 209938165 - 210171315 on Build GRCh38
UniGene ID
Hs.658866
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146261.2 NP_001139733.1 7 1352
NM_001146262.2 NP_001139734.1 6 1231
NM_001146264.2 NP_001139736.1 7 1352
NM_001256006.1 NP_001242935.1 7 1436
NM_153262.3 NP_694994.2 6 1231
XM_006711262.3 XP_006711325.1 5 2410
XM_017000931.1 XP_016856420.1 5 2410
XM_017000932.1 XP_016856421.1 7 1426
XM_017000933.1 XP_016856422.1 7 1289
XM_017000934.1 XP_016856423.1 4 1085
XM_017000935.1 XP_016856424.1 5 968
GenBank mRNA AB102948.1 - 5 1140
AJ617623.1 - 6 1174
AJ617624.1 - 6 1317
AJ617626.1 - 3 907
AJ617627.1 - 2 666
AK091517.1 - 6 1154
BC130323.1 - 7 1285 40146354
BC144154.1 - 7 1285
BC144155.1 - 6 1164
BC144157.1 - 7 1285

View Full Product Details