Gene Transcripts

Gene Symbol
KLHL34
Entrez Gene ID
257240
Gene Name
kelch like family member 34
Gene Aliases
Chromosome Location
Chr. X: 21655349 - 21658387 on Build GRCh38
UniGene ID
Hs.448572
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153270.2 NP_695002.1 1 2114
GenBank mRNA AK092279.1 - 1 2057
BC109031.2 - 1 2026 40027951

View Full Product Details