Gene Transcripts

Gene Symbol
MAMSTR
Entrez Gene ID
284358
Gene Name
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases
MASTR
Chromosome Location
Chr. 19: 48705718 - 48719898 on Build GRCh38
UniGene ID
Hs.191815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130915.1 NP_001124387.1 8, 9 972
NM_001297753.1 NP_001284682.1 5, 6 1085
NM_182574.2 NP_872380.1 6, 7 1216
XM_011526807.2 XP_011525109.1 6, 7 1221
XM_011526808.2 XP_011525110.1 8, 9 1340
XM_011526809.2 XP_011525111.1 5, 6 1019
XM_017026640.1 XP_016882129.1 7, 8 1740
XM_017026641.1 XP_016882130.1 7, 8 1701
XM_017026643.1 XP_016882132.1 7, 8 1581
XM_017026644.1 XP_016882133.1 7, 8 1551
XM_017026645.1 XP_016882134.1 4, 5 986
GenBank mRNA AK093389.1 - 6, 7 1216
AK131427.1 - 8, 9 979
BC105056.1 - 4, 5 640 8144059
BC105058.1 - 4, 5 640 8144061
BC106047.1 - 5, 6 1085 5786639
BC143443.1 - 3, 4 445 9051951
BX280379.1 - 1, 2 166

View Full Product Details