Gene Transcripts
- Gene Symbol
- KLK9
- Entrez Gene ID
- 284366
- Gene Name
- kallikrein related peptidase 9
- Gene Aliases
- KLK-L3, KLKL3
- Chromosome Location
- Chr. 19: 51002513 - 51009634 on Build GRCh38
- UniGene ID
- Hs.448942
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012315.1 |
NP_036447.1
|
5 |
1382 |
|
- Gene Symbol
- KLK8
- Entrez Gene ID
- 11202
- Gene Name
- kallikrein related peptidase 8
- Gene Aliases
- HNP, NP, NRPN, PRSS19, TADG14
- Chromosome Location
- Chr. 19: 50996008 - 51001702 on Build GRCh38
- UniGene ID
- Hs.104570
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC040887.1 |
- |
1 |
143 |
|
View Full Product Details