Gene Transcripts
- Gene Symbol
- PKD1L3
- Entrez Gene ID
- 342372
- Gene Name
- polycystin 1 like 3, transient receptor potential channel interacting
- Gene Aliases
- Chromosome Location
- Chr. 16: 71929538 - 72000401 on Build GRCh38
- UniGene ID
- Hs.675286
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181536.1 |
NP_853514.1
|
28 |
4888 |
|
GenBank mRNA |
AY164485.1 |
- |
28 |
4888 |
|
View Full Product Details