Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039361.4 | NP_001034450.3 | 4 | 1335 | |
GenBank mRNA | AL049682.1 | - | 4 | 1335 | |
BC112208.1 | - | 4 | 1331 | 8327676 | |
BC112210.1 | - | 4 | 1331 | 8327678 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291381.1 | NP_001278310.1 | 4 | 1328 |