Gene Transcripts

Gene Symbol
OR6F1
Entrez Gene ID
343169
Gene Name
olfactory receptor family 6 subfamily F member 1
Gene Aliases
OR1-34, OR1-38, OST731
Chromosome Location
Chr. 1: 247711829 - 247712755 on Build GRCh38
UniGene ID
Hs.591529
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005286.1 NP_001005286.1 1 273
GenBank mRNA BC137142.1 - 1 321
BC137143.1 - 1 321 9021140

View Full Product Details