Gene Transcripts
- Gene Symbol
- RILPL1
- Entrez Gene ID
- 353116
- Gene Name
- Rab interacting lysosomal protein like 1
- Gene Aliases
- GOSPEL, RLP1
- Chromosome Location
- Chr. 12: 123471073 - 123533718 on Build GRCh38
- UniGene ID
- Hs.530315
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319243.1 |
NP_001306172.1
|
5, 6 |
1204 |
|
NM_001319244.1 |
NP_001306173.1
|
5, 6 |
753 |
|
NM_001319302.1 |
NP_001306231.1
|
6, 7 |
1703 |
|
NM_178314.4 |
NP_847884.2
|
5, 6 |
1204 |
|
XM_006719372.2 |
XP_006719435.1
|
5, 6 |
1198 |
|
XM_006719373.3 |
XP_006719436.1
|
4, 5 |
551 |
|
XM_011538282.2 |
XP_011536584.1
|
5, 6 |
1239 |
|
XM_011538284.2 |
XP_011536586.1
|
4, 5 |
770 |
|
XM_011538285.2 |
XP_011536587.1
|
4, 5 |
531 |
|
GenBank mRNA |
AK096697.1 |
- |
6, 7 |
1046 |
|
BC080626.1 |
- |
5, 6 |
753 |
6339887 |
BC089444.1 |
- |
5, 6 |
1006 |
30559974 |
View Full Product Details