Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304506.1 | NP_001291435.1 | 5 | 568 | |
NM_019110.4 | NP_061983.2 | 5 | 1233 | ||
XM_005249095.3 | XP_005249152.1 | 7 | 1277 | ||
XM_005249097.3 | XP_005249154.1 | 7 | 973 | ||
XM_005249098.3 | XP_005249155.1 | 5 | 891 | ||
XM_005249100.4 | XP_005249157.2 | 5 | 624 | ||
XM_006715095.3 | XP_006715158.1 | 6 | 750 | ||
XM_011514588.2 | XP_011512890.1 | 5 | 1133 | ||
XM_011514589.2 | XP_011512891.1 | 5 | 689 | ||
XM_017010844.1 | XP_016866333.1 | 7 | 1928 | ||
GenBank mRNA | AK302017.1 | - | 5 | 549 | |
AK315783.1 | - | 5 | 1203 | ||
AY781778.1 | - | 5 | 1203 | ||
BC014031.2 | - | 5 | 1161 | 4336904 | |
KU178905.1 | - | 5 | 917 | ||
KU178906.1 | - | 4 | 806 |