Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039492.2 | NP_001034581.1 | 7 | 967 | |
NM_001318894.1 | NP_001305823.1 | 6 | 1213 | ||
NM_001318895.1 | NP_001305824.1 | 7 | 1201 | ||
NM_001318896.1 | NP_001305825.1 | 6 | 916 | ||
NM_001318897.1 | NP_001305826.1 | 5 | 967 | ||
NM_001318898.1 | NP_001305827.1 | 6 | 1026 | ||
NM_001318899.1 | NP_001305828.1 | 8 | 1089 | ||
NM_001450.3 | NP_001441.4 | 6 | 1150 | ||
NM_201555.1 | NP_963849.1 | 8 | 1322 | ||
NM_201557.3 | NP_963851.2 | 6 | 1142 | ||
XM_011510798.2 | XP_011509100.1 | 7 | 949 | ||
GenBank mRNA | AB158503.1 | - | 5 | 689 | |
AK097756.1 | - | 1 | 1553 | ||
AK313841.1 | - | 7 | 999 | ||
AL521653.3 | - | 6 | 843 | ||
AY634685.1 | - | 5 | 743 | ||
BC012742.2 | - | 8 | 1322 | 3840517 | |
BC014397.1 | - | 8 | 1322 | 3546686 | |
BC036085.1 | - | 6 | 797 | ||
BC093049.1 | - | 6 | 908 | 30374919 | |
BI092736.1 | - | 3 | 260 | ||
BT006960.1 | - | 5 | 743 | ||
BX340370.2 | - | 2 | 110 | ||
CR457437.1 | - | 5 | 743 | ||
CR936682.1 | - | 6 | 1213 | ||
CR936799.1 | - | 6 | 6194 | ||
DQ266432.1 | - | 7 | 1201 | ||
DQ307067.1 | - | 6 | 1080 | ||
JQ313902.1 | - | 5 | 713 | ||
L42176.1 | - | 7 | 881 | ||
U29332.1 | - | 6 | 864 | ||
U60117.1 | - | 3 | 366 |