Gene Transcripts
- Gene Symbol
- NHLRC3
- Entrez Gene ID
- 387921
- Gene Name
- NHL repeat containing 3
- Gene Aliases
- Chromosome Location
- Chr. 13: 39038311 - 39050109 on Build GRCh38
- UniGene ID
- Hs.507783
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012754.3 |
NP_001012772.1
|
6, 7 |
1128 |
|
NM_001017370.2 |
NP_001017370.1
|
5, 6 |
927 |
|
NR_073109.1 |
|
6, 7 |
870 |
|
GenBank mRNA |
AK299198.1 |
- |
6, 7 |
1041 |
|
AK300260.1 |
- |
5, 6 |
840 |
|
BC052492.1 |
- |
6, 7 |
808 |
|
BC140877.1 |
- |
6, 7 |
1054 |
9021745 |
BC140878.1 |
- |
6, 7 |
1054 |
9021746 |
BX647872.1 |
- |
5, 6 |
817 |
|
BX648122.1 |
- |
6, 7 |
1080 |
|
CR749318.1 |
- |
5, 6 |
849 |
|
View Full Product Details