Gene Transcripts

Gene Symbol
SERP2
Entrez Gene ID
387923
Gene Name
stress-associated endoplasmic reticulum protein family member 2
Gene Aliases
C13orf21, bA269C23.1
Chromosome Location
Chr. 13: 44373842 - 44397714 on Build GRCh38
UniGene ID
Hs.377972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010897.1 NP_001010897.1 3 748
GenBank mRNA AK123212.1 - 1 1548
BC029067.1 - 3 748 5197847

View Full Product Details