Gene Transcripts

Gene Symbol
KRTDAP
Entrez Gene ID
388533
Gene Name
keratinocyte differentiation associated protein
Gene Aliases
KDAP, UNQ467
Chromosome Location
Chr. 19: 35487324 - 35490531 on Build GRCh38
UniGene ID
Hs.112457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244847.1 NP_001231776.1 2 196
NM_207392.2 NP_997275.1 2 196
GenBank mRNA AJ293408.1 - 2 157
AY359007.1 - 2 119
BC130501.1 - 2 196 40146532
W95920.1 - 2 105

View Full Product Details