Gene Transcripts

Gene Symbol
OR5B12
Entrez Gene ID
390191
Gene Name
olfactory receptor family 5 subfamily B member 12
Gene Aliases
OR11-241, OR5B12P, OR5B16, OST743
Chromosome Location
Chr. 11: 58439120 - 58440173 on Build GRCh38
UniGene ID
Hs.553754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004733.2 NP_001004733.1 1 495
GenBank mRNA BC136947.1 - 1 495
BC136948.1 - 1 495 9020940

View Full Product Details