Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000344.3 | NP_000335.1 | 7 | 946 | |
NM_001297715.1 | NP_001284644.1 | 7 | 946 | ||
NM_022874.2 | NP_075012.1 | 6 | 850 | ||
XM_011543596.1 | XP_011541898.1 | 7 | 825 | ||
XM_011543597.1 | XP_011541899.1 | 6 | 630 | ||
XM_011543598.2 | XP_011541900.1 | 5 | 545 | ||
XM_017009786.1 | XP_016865275.1 | 6 | 729 | ||
GenBank mRNA | AK298203.1 | - | 6 | 630 | |
BC062723.1 | - | 7 | 802 | 4250429 | |
JQ657798.1 | - | 4 | 439 | ||
JQ657799.1 | - | 4 | 439 | ||
JQ657800.1 | - | 4 | 334 | ||
JQ657802.1 | - | 4 | 439 | ||
JQ657803.1 | - | 3 | 238 | ||
JQ690867.1 | - | 4 | 334 | ||
JQ690868.1 | - | 3 | 238 | ||
JQ745297.1 | - | 3 | 181 | ||
U18423.1 | - | 7 | 816 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017411.3 | NP_059107.1 | 7 | 946 | |
NM_022875.2 | NP_075013.1 | 7 | 946 | ||
NM_022876.2 | NP_075014.1 | 6 | 850 | ||
NM_022877.2 | NP_075015.1 | 6 | 850 | ||
XM_011543599.1 | XP_011541901.1 | 7 | 825 | ||
XM_011543600.2 | XP_011541902.1 | 6 | 630 | ||
XM_011543601.1 | XP_011541903.1 | 6 | 630 | ||
XM_011543602.2 | XP_011541904.1 | 5 | 546 | ||
XM_011543603.2 | XP_011541905.1 | 5 | 546 | ||
XM_017009787.1 | XP_016865276.1 | 7 | 825 | ||
GenBank mRNA | AK289669.1 | - | 7 | 829 | |
BC000908.2 | - | 7 | 815 | 2985046 | |
BC015308.1 | - | 7 | 813 | 4427000 | |
BC070242.1 | - | 7 | 792 | 4673318 | |
CN272420.1 | - | 5 | 649 | ||
JQ690861.1 | - | 4 | 334 | ||
JQ690862.1 | - | 4 | 334 | ||
JQ690863.1 | - | 3 | 238 | ||
JQ690864.1 | - | 4 | 382 | ||
JQ690865.1 | - | 4 | 439 | ||
JQ690866.1 | - | 3 | 238 | ||
U21914.1 | - | 7 | 780 |