Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018713.2 | NP_061183.2 | 4 | 2210 | |
NR_046437.1 | 4 | 2248 | |||
XM_006711437.3 | XP_006711500.2 | 4 | 2546 | ||
XM_017001684.1 | XP_016857173.1 | 4 | 1797 | ||
GenBank mRNA | AK090806.1 | - | 4 | 2248 | |
AL359609.1 | - | 2 | 1816 | ||
BC036078.1 | - | 4 | 1770 | ||
BG564297.1 | - | 1 | 225 | ||
CB999545.1 | - | 1 | 224 |