Gene Transcripts

Gene Symbol
TMIE
Entrez Gene ID
259236
Gene Name
transmembrane inner ear
Gene Aliases
DFNB6
Chromosome Location
Chr. 3: 46693800 - 46710923 on Build GRCh38
UniGene ID
Hs.185777
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147196.2 NP_671729.2 4 797
XM_006713097.3 XP_006713160.1 4 1368
GenBank mRNA AY081842.1 - 4 794
BC126258.1 - 4 757 8991974
BC126260.1 - 4 757 8991976

View Full Product Details