Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
11330
Gene Name
chymotrypsin C
Gene Aliases
CLCR, ELA4
Chromosome Location
Chr. 1: 15438442 - 15449247 on Build GRCh38
UniGene ID
Hs.631869
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007272.2 NP_009203.2 7, 8 808
XM_011540550.1 XP_011538852.1 6, 7 662
GenBank mRNA AK289447.1 - 7, 8 807
BC015118.1 - 7, 8 814 3950170
BC059779.1 - 6, 7 641
CR749296.1 - 4, 5 472
S82198.1 - 7, 8 785
Y13697.1 - 6, 7 737

View Full Product Details