Gene Transcripts

Gene Symbol
SIRT3
Entrez Gene ID
23410
Gene Name
sirtuin 3
Gene Aliases
SIR2L3
Chromosome Location
Chr. 11: 215030 - 236950 on Build GRCh38
UniGene ID
Hs.716456
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005252835.1 XP_005252892.1 6, 7 1275
XM_011519956.1 XP_011518258.1 6, 7 2067
XM_011519957.1 XP_011518259.1 6, 7 1896
XM_017017428.1 XP_016872917.1 6, 7 1121
XM_017017429.1 XP_016872918.1 6, 7 1295
XM_017017431.1 XP_016872920.1 6, 7 1020
GenBank mRNA AK074992.1 - 4, 5 2061
AK301558.1 - 5, 6 1075
AK301979.1 - 6, 7 1263

View Full Product Details