Gene Transcripts

Gene Symbol
UQCRHL
Entrez Gene ID
440567
Gene Name
ubiquinol-cytochrome c reductase hinge protein like
Gene Aliases
Chromosome Location
Chr. 1: 15807162 - 15807699 on Build GRCh38
UniGene ID
Hs.568229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001089591.1 NP_001083060.1 1 486

View Full Product Details