Gene Transcripts

Gene Symbol
OR5B3
Entrez Gene ID
441608
Gene Name
olfactory receptor family 5 subfamily B member 3
Gene Aliases
OR11-239, OR5B13, OST129
Chromosome Location
Chr. 11: 58402465 - 58403409 on Build GRCh38
UniGene ID
Hs.553826
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005469.1 NP_001005469.1 1 49

View Full Product Details