Gene Transcripts

Gene Symbol
SPIN2B
Entrez Gene ID
474343
Gene Name
spindlin family member 2B
Gene Aliases
SPIN-2, SPIN-2B, SPIN2_duplicate, TDRD26, dJ323P24.2
Chromosome Location
Chr. X: 57112142 - 57121905 on Build GRCh38
UniGene ID
Hs.460717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006681.1 NP_001006682.1 2 845
NM_001006682.1 NP_001006683.1 2 793
NM_001006683.1 NP_001006684.1 2 790
XM_005262008.4 XP_005262065.2 2 639
XM_005262009.4 XP_005262066.2 2 628
XM_005262010.1 XP_005262067.1 2 850
XM_011530788.2 XP_011529090.1 2 640
XM_017029552.1 XP_016885041.1 2 637
GenBank mRNA AF356353.1 - 2 662
BC000044.2 - 2 738 3504291
BC071974.1 - 2 777 6729986

Gene Symbol
SPIN2A
Entrez Gene ID
54466
Gene Name
spindlin family member 2A
Gene Aliases
DXF34, SPIN2, TDRD25, dJ323P24.1
Chromosome Location
Chr. X: 57134259 - 57138688 on Build GRCh38
UniGene ID
Hs.460725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019003.3 NP_061876.3 2 905
XM_005262016.4 XP_005262073.3 2 558
XM_005262017.4 XP_005262074.3 2 558
XM_005262019.3 XP_005262076.1 2 2020
XM_017029598.1 XP_016885087.1 2 1965
GenBank mRNA BC071694.1 - 2 905
Y09858.1 - 1 949

View Full Product Details