Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000154.1 | NP_000145.1 | 2 | 400 | |
GenBank mRNA | AK130064.1 | - | 690 | ||
AK303748.1 | - | 2 | 374 | ||
AK303832.1 | - | 2 | 437 | ||
AK310203.1 | - | 2 | 393 | ||
AK314890.1 | - | 2 | 360 | ||
AK316181.1 | - | 2 | 358 | ||
BC001166.1 | - | 2 | 381 | 3501788 | |
BT007005.1 | - | 2 | 337 | ||
EU794610.1 | - | 2 | 365 | ||
U26401.1 | - | 2 | 365 |