Gene Transcripts

Gene Symbol
LINC00520
Entrez Gene ID
645687
Gene Name
long intergenic non-protein coding RNA 520
Gene Aliases
C14orf34
Chromosome Location
Chr. 14: 55781135 - 55796674 on Build GRCh38
UniGene ID
Hs.569345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026796.1 4 683
NR_026797.1 3 621
GenBank mRNA BU171486.1 - 4 683
BU178088.1 - 3 633

View Full Product Details